missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium plate, 6.4mm (0.25 in.) thick, 100x100mm (3.9x3.9in), 99.2% (metals basis excluding Hf)

Manufacturer:  Alfa Aesar™ 44261KF

 View more versions of this product

Catalog No. AA44261KF



Specifications

Specifications

Zirconium plate
1 pc.
7440-67-7
Zr
and compounds
[Zr]
Odorless
23995
Plate
≈416g/100x100mm
6.4mm (0.25 in.) thick, 100x100mm (3.9x3.9 in.), Temper: annealed
(metals basis excluding Hf)
MFCD00011303
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zirconium
91.224
CHEBI:33342
99.2%

Chemical Identifiers

7440-67-7
91.224
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
zirconium
Zr
MFCD00011303
and compounds
CHEBI:33342
[Zr]
Safety and Handling

Safety and Handling

EINECSNumber : 231-176-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents