missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zirconium plate, 6.4mm (0.25in) thick, 100x100mm (3.9x3.9in), 99.2% (metals basis excluding Hf), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 044261KJ
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zirconium plate | |
7440-67-7 | |
Zr | |
5 Pc. | |
QCWXUUIWCKQGHC-UHFFFAOYSA-N | |
zirconium | |
23995 | |
99.2% | |
≈416g/100x100mm |
6.4mm (0.25 in.) thick, 100x100mm (3.9x3.9 in.), Temper: annealed | |
(metals basis excluding Hf) | |
MFCD00011303 MFCD00148878 | |
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil | |
[Zr] | |
91.22 | |
CHEBI:33342 | |
Odorless | |
Plate |
Chemical Identifiers
7440-67-7 | |
91.22 | |
QCWXUUIWCKQGHC-UHFFFAOYSA-N | |
23995 | |
zirconium |
Zr | |
MFCD00011303 MFCD00148878 | |
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil | |
CHEBI:33342 | |
[Zr] |
Safety and Handling
EINECSNumber : 231-176-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only