Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium plate, 3.175mm (0.125 in.) thick, 99.2% (metals basis excluding Hf), Hf 4.5% max

Chemical Identifiers

Chemical Name or Material Zirconium plate
CAS 7440-67-7
Name Note 3.175mm (0.125 in.) thick
MDL Number MFCD00011303
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA10593GH

  SDS
alfa aesar™
10593GH
100x100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Zirconium plate
TSCA TSCA
Name Note 3.175mm (0.125 in.) thick
Physical Form Plate
Assay 99.2%
Assay Percent Notes (metals basis excluding Hf), Hf 4.5% max.
MDL Number MFCD00011303
CAS 7440-67-7
Odor Odorless
Weight ≈206.6g/100x100mm
Quantity 100x100mm