missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium, plasma standard solution, Specpure™ Zr 10,000μg/mL, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 014386AE

 View more versions of this product

Catalog No. AA14386AE



Description

Description

Zirconium plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zirconium plasma standard solution
Zr 10000μg/mL
MFCD00011303
Specpure™
Liquid
ZrCl2O in 5% HCl
100 mL
UN1789
Miscible with water

Chemical Identifiers

MFCD00011303
Safety and Handling

Safety and Handling

GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HYDROCHLORIC ACID

EINECSNumber : 231-595-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only