missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium oxide, yttria stabilized, polymeric precursor

As a solid electrolyte and thermal barrier coating

Manufacturer:  Alfa Aesar™ 4421103

 View more versions of this product

Catalog No. AA4421103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Zirconium oxide
1314-23-4
MFCD00011310
pcs filler
MCMNRKCIXSYSNV-UHFFFAOYSA-N
dioxozirconium
62395
1g
Yttria stabilized, polymeric precursor
O2Zr
UN1993
Soluble in hexane, toluene, alcohol
O=[Zr]=O
123.222
Liquid

Chemical Identifiers

1314-23-4
123.222
MCMNRKCIXSYSNV-UHFFFAOYSA-N
62395
O=[Zr]=O
O2Zr
MFCD00011310
pcs filler
dioxozirconium
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 215-227-2

TSCA : No

Recommended Storage : Ambient temperatures

Documents