Please login to your online account to display your discounted pricing

Zirconium oxide, Yttria stabilized, Grinding Pieces, 5mm diameter, Alfa Aesar™

Specifications: Yttrium oxide (mole) 3%, Hardness 12, Total impuroty level <0.1

$179.31 - $578.59

Chemical Identifiers

Chemical Name or Material Zirconium oxide
MDL Number MFCD00145546
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4328530

  SDS
alfa aesar
4328530
250g Each for $179.31

AA43285A1

  SDS
alfa aesar
43285A1
1kg Each for $578.59
Description & Specifications

Specifications

Quantity 250g
Color Off-white
Odor Odorless
Appearance Off-white chunks
MDL Number MFCD00145546
Chemical Name or Material Zirconium oxide