Please login to your online account to display your discounted pricing

Zirconium Oxide, Catalyst Support, Tungstated, Alfa Aesar™

$139.62 - $1,553.14

Chemical Identifiers

Chemical Name or Material Zirconium Oxide
MDL Number MFCD00168096
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4559722

 
alfa aesar
4559722
100g Each for $139.62

AA4559736

 
alfa aesar
4559736
500g Each for $444.85

AA45597A4

 
alfa aesar
45597A4
2.5kg Each for $1,553.14
Description & Specifications

Specifications

Quantity 100g
Chemical Name or Material Zirconium Oxide
Particle Size 3mm
MDL Number MFCD00168096
Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.