Please login to your online account to display your discounted pricing

Zirconium Oxide, Catalyst Support, Sulfated, Alfa Aesar™

$145.15 - $2,353.55

Chemical Identifiers

Chemical Name or Material Zirconium Oxide
MDL Number MFCD01863639
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4560022

  SDS
alfa aesar
4560022
100g Each for $145.15

AA4560036

  SDS
alfa aesar
4560036
500g Each for $541.82

AA45600A4

  SDS
alfa aesar
45600A4
2.5kg Each for $2,353.55
Description & Specifications

Specifications

Quantity 100g
Color White
Chemical Name or Material Zirconium Oxide
Odor Odorless
Particle Size 3mm
Appearance White pellets
MDL Number MFCD01863639
Health Hazard 1 H315-H319: Causes skin irritation. Causes serious eye irritation.
Safety and Handling

H315-H319: Causes skin irritation. Causes serious eye irritation.