missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium nitride, 99.5% (metals basis excluding Hf), Hf >3%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1213814

 View more versions of this product

Catalog No. AA1213814



Specifications

Specifications

Zirconium nitride
99.5% (metals basis excluding Hf)
MFCD00049642
ZVWKZXLXHLZXLS-UHFFFAOYSA-N
azanylidynezirconium
94359
-325 Mesh Powder
25g
25658-42-8
NZr
zirconium nitride
N#[Zr]
105.231
105.23
7.09g/mL

Chemical Identifiers

25658-42-8
105.231
ZVWKZXLXHLZXLS-UHFFFAOYSA-N
94359
N#[Zr]
NZr
MFCD00049642
zirconium nitride
azanylidynezirconium
Safety and Handling

Safety and Handling

EINECSNumber : 247-166-2

Documents