Please login to your online account to display your discounted pricing

Zirconium nitride, 99.5% (metals basis excluding Hf), Hf >3%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Zirconium nitride
CAS 25658-42-8
Physical Form -325 Mesh Powder
Assay Percent Range 99.5% (metals basis excluding Hf)
Molecular Formula NZr
Formula Weight 105.23
MDL Number MFCD00049642
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1213814

  SDS
alfa aesar
1213814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1213822

  SDS
alfa aesar
1213822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 105.23
Quantity 25g
EINECS Number 247-166-2
Physical Form -325 Mesh Powder
MDL Number MFCD00049642
Chemical Name or Material Zirconium nitride
Density 7.09g/mL
Assay Percent Range 99.5% (metals basis excluding Hf)
Molecular Formula NZr
CAS 25658-42-8