Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium(IV) tert-butoxide, 99% (metals basis)

$893.51 - $893.51

Chemical Identifiers

Chemical Name or Material Zirconium(IV) tert-butoxide
CAS 2081-12-1
Linear Formula Zr[OC(CH3)3]4
Formula Weight 383.68
MDL Number MFCD00075085
Beilstein 3681870
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3954703

  SDS
alfa aesar™
3954703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3954706

  SDS
alfa aesar™
3954706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3954714

  SDS
alfa aesar™
3954714
25g Each for $893.51
Description & Specifications

Specifications

Formula Weight 383.68
Physical Form Liquid
Color Yellow
Quantity 1g
Sensitivity Moisture & Light Sensitive
Assay Percent Notes (metals basis)
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 2081-12-1
Density 0.985g/mL
Linear Formula Zr[OC(CH3)3]4
Chemical Name or Material Zirconium(IV) tert-butoxide
Assay 99%
MDL Number MFCD00075085
Boiling Point 90°C (5mmHg)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Flash Point 85° (185°F)
Beilstein 3681870
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.