missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium(IV) oxide, yttria stabilized, 99.95% (metals basis excluding Hf)

Manufacturer:  Alfa Aesar™ 4556222

 View more versions of this product

Catalog No. AA4556222



Specifications

Specifications

Zirconium (IV) oxide
1314-23-4
O2Zr
14,10180
MCMNRKCIXSYSNV-UHFFFAOYSA-N
dioxozirconium
62395
99.95%
−325 Mesh
~5000°C
~2700°C
Yttria stabilized
(metals basis excluding Hf)
MFCD00145546
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
O=[Zr]=O
123.222
Powder
ZrO2 + 8% Y2O3
Odorless
White
100g

Chemical Identifiers

1314-23-4
123.222
MCMNRKCIXSYSNV-UHFFFAOYSA-N
62395
O=[Zr]=O
O2Zr
MFCD00145546
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
dioxozirconium
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents