missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium(IV) oxide, titania stablized

Manufacturer:  Alfa Aesar™ 4512606

 View more versions of this product

Catalog No. AA4512606



Specifications

Specifications

Zirconium (IV) oxide
1314-23-4
MFCD00011310
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
MCMNRKCIXSYSNV-UHFFFAOYSA-N
dioxozirconium
62395
5.4g/mL
Black
5g
Titania stablized
O2Zr
14,10180
ZrO2:TiO2, 90:10 wt%
O=[Zr]=O
123.222
Tablets
Odorless
~2500°C
≈1g Tablets

Chemical Identifiers

1314-23-4
123.222
MCMNRKCIXSYSNV-UHFFFAOYSA-N
62395
O=[Zr]=O
O2Zr
MFCD00011310
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
dioxozirconium
Safety and Handling

Safety and Handling

RTECSNumber : ZH8800000

TSCA : TSCA

SDS Documents