missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zirconium(IV) oxide, calcia stabilized, 99.4% (metals basis excluding Hf), Thermo Scientific Chemicals
$74.40 - $588.03
Chemical Identifiers
CAS | 1314-23-4 |
---|---|
Molecular Formula | O2Zr |
Molecular Weight (g/mol) | 123.22 |
MDL Number | MFCD00011310 MFCD01868884 |
InChI Key | MCMNRKCIXSYSNV-UHFFFAOYSA-N |
Synonym | zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler |
PubChem CID | 62395 |
IUPAC Name | dioxozirconium |
SMILES | O=[Zr]=O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-23-4 | |
123.22 | |
MCMNRKCIXSYSNV-UHFFFAOYSA-N | |
62395 | |
O=[Zr]=O |
O2Zr | |
MFCD00011310 MFCD01868884 | |
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler | |
dioxozirconium |
Specifications
∼5000°C | |
1314-23-4 | |
50 g | |
MFCD00011310 MFCD01868884 | |
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler | |
O=[Zr]=O | |
123.22 | |
123.22 | |
ZrO2, 4% CaO | |
Zirconium(IV) oxide, Calcia stabilized |
∼2700°C | |
Powder | |
O2Zr | |
14,10180 | |
MCMNRKCIXSYSNV-UHFFFAOYSA-N | |
dioxozirconium | |
62395 | |
99.4% | |
Odorless | |
(metals basis excluding Hf) |
Safety and Handling
EINECSNumber : 215-227-2
RTECSNumber : ZH8800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only