Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium(IV) oxide, Aerosol Refractory Paint, 99% (including 4% CaO)

Chemical Identifiers

Chemical Name or Material Zirconium(IV) oxide
CAS 1314-23-4
Name Note Aerosol Refractory Paint
MDL Number MFCD00011310
UN Number UN1950
Merck Index 14,10180
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA404800CL

  SDS
alfa aesar™
404800CL
5 x 13 oz. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P210-P251-P211-P280-P305+P351+P338-P410+P412
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Pressurized container: Do not pierce or burn, even after use.
Do not spray on an open flame or other ignition source.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Protect from sunlight. Do no expose to temperatures exceeding 50 celcius degress.
Health Hazard 1 Danger
UN Number UN1950
Chemical Name or Material Zirconium(IV) oxide
TSCA TSCA
Color Cream
Merck Index 14,10180
DOT Information Transport Hazard Class: 2.1; Packaging Group: ; Proper Shipping Name: AEROSOLS
Packaging Aerosol Spray Can
Assay 99%
CAS 1314-23-4
Quantity 5 x 13 oz.
Name Note Aerosol Refractory Paint
Health Hazard 2 GHS H Statement
H222-H319
Extremely flammable aerosol.
Causes serious eye irritation.
Assay Percent Notes including 4% CaO
MDL Number MFCD00011310
Safety and Handling

Danger

GHS P Statement
P210-P251-P211-P280-P305+P351+P338-P410+P412
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Pressurized container: Do not pierce or burn, even after use.
Do not spray on an open flame or other ignition source.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Protect from sunlight. Do no expose to temperatures exceeding 50 celcius degress.

GHS H Statement
H222-H319
Extremely flammable aerosol.
Causes serious eye irritation.