missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium(IV) Oxide, ≥99% (Metals Basis excluding Hf), HfO2 2%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1139530

 View more versions of this product

Catalog No. AA1139530



Specifications

Specifications

Zirconium(IV) Oxide
≥99%, metal basis
MFCD00011310
MCMNRKCIXSYSNV-UHFFFAOYSA-N
dioxozirconium
62395
-325 Mesh Powder
250g
1314-23-4
O2Zr
pcs filler
O=[Zr]=O
123.222
123.22
5.89

Chemical Identifiers

1314-23-4
123.222
MCMNRKCIXSYSNV-UHFFFAOYSA-N
62395
O=[Zr]=O
O2Zr
MFCD00011310
pcs filler
dioxozirconium
Documents