Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium(IV) oxide, 99.8% (metals basis excluding Hf)

Catalyst support, chemical sensors

Chemical Identifiers

Chemical Name or Material Zirconium(IV) oxide
CAS 1314-23-4
Formula Weight 123.22
MDL Number MFCD00011310
Merck Index 14,10180
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4045904

  SDS
alfa aesar™
4045904
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4045909

  SDS
alfa aesar™
4045909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 123.22
Chemical Name or Material Zirconium(IV) oxide
TSCA TSCA
Mesh Size −325 Mesh
Odor Odorless
Assay 99.8%
Assay Percent Notes (metals basis excluding Hf)
Surface Area 100 to 200m2/g
MDL Number MFCD00011310
CAS 1314-23-4
Merck Index 14,10180
Boiling Point ≈5000°C
Color White
Melting Point ≈2700°C
Quantity 2g
Density 5.89g/mL
RTECS Number ZH8800000