Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium(IV) oxide, 99.5% (metals basis excluding Hf), Hf <100ppm

Chemical Identifiers

Chemical Name or Material Zirconium(IV) oxide
CAS 1314-23-4
Formula Weight 123.22
MDL Number MFCD00011310
Merck Index 14,10180
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4014022

  SDS
alfa aesar™
4014022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4014036

  SDS
alfa aesar™
4014036
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 123.22
Chemical Name or Material Zirconium(IV) oxide
TSCA TSCA
Odor Odorless
Particle Size 1 Micron APS
Physical Form Powder
Assay 99.5%
Assay Percent Notes (metals basis excluding Hf), Hf <100ppm
MDL Number MFCD00011310
CAS 1314-23-4
Merck Index 14,10180
Boiling Point ≈5000°C
Color White
Melting Point ≈2700°C
Quantity 100g
Density 5.89g/mL
RTECS Number ZH8800000