Learn More
Zirconium(IV) oxide, 98.5%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 190522500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4300.0°C | |
2700.0°C | |
1314-23-4 | |
Powder | |
98% min. (ZrO2+ HfO2) | |
ZrO2 | |
15, 10380 | |
Solubility in water: insoluble <1mg/l (20°C). Other solubilities: slightly soluble in hcl and hno3, slowly soluble in hf, disolves on heating with a mixt of 2 parts, h2so4 and 1 part h2o | |
O=[Zr]=O | |
123.22 | |
123.22 | |
Glass bottle | |
5.6000g/mL |
Zirconium(IV) oxide | |
98.5% | |
White | |
250 g | |
O2Zr | |
MFCD00011310 MFCD01868884 | |
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler | |
MCMNRKCIXSYSNV-UHFFFAOYSA-N | |
dioxozirconium | |
62395 | |
98.5% | |
5.6 |
Chemical Identifiers
1314-23-4 | |
123.22 | |
MCMNRKCIXSYSNV-UHFFFAOYSA-N | |
62395 | |
O=[Zr]=O |
O2Zr | |
MFCD00011310 MFCD01868884 | |
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler | |
dioxozirconium |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 215-227-2
RTECSNumber : ZH8800000
TSCA : TSCA
RUO – Research Use Only