Learn More
Zirconium(IV) oxide 2-ethylhexanoate, in mineral spirits (≈6% Zr), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03954336
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zirconium(IV) oxide 2-ethylhexanoate | |
22464-99-9 | |
C8H16O2Zr | |
500 g | |
zirconium 2-ethylhexanoate | |
CCCCC(CC)C(=O)O.[Zr] | |
235.438 | |
≈6% Zr |
In mineral spirits | |
40°C (104°F) | |
MFCD00072684 | |
UN1268 | |
HBHXRPHNNKAWQL-UHFFFAOYSA-N | |
2-ethylhexanoic acid;zirconium | |
73555628 | |
Liquid |
Chemical Identifiers
22464-99-9 | |
235.438 | |
HBHXRPHNNKAWQL-UHFFFAOYSA-N | |
73555628 | |
CCCCC(CC)C(=O)O.[Zr] |
C8H16O2Zr | |
MFCD00072684 | |
zirconium 2-ethylhexanoate | |
2-ethylhexanoic acid;zirconium |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P314-P330-P370+P378q-P501c
H226-H302-H336-H372
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.
TSCA : Yes
RUO – Research Use Only