missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium(IV) n-propoxide, 70% w/w in n-propanol, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 4477018

 View more versions of this product

Catalog No. AA4477018



Description

Description

  • Packaged under Argon
Specifications

Specifications

Zirconium (IV) n-propoxide
28°C (83°F)
1.4510
50g
C12H28O4Zr
UN1993
70% w/w in n-propanol
CCC[O-].CCC[O-].CCC[O-].CCC[O-].[Zr+4]
327.576
327.56
1.044g/mL
ChemSeal™ bottles
Moisture Sensitive
23519-77-9
MFCD00015307
zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt
XPGAWFIWCWKDDL-UHFFFAOYSA-N
propan-1-olate;zirconium(4+)
90139
Liquid

Chemical Identifiers

23519-77-9
327.576
XPGAWFIWCWKDDL-UHFFFAOYSA-N
90139
CCC[O-].CCC[O-].CCC[O-].CCC[O-].[Zr+4]
C12H28O4Zr
MFCD00015307
zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt
propan-1-olate;zirconium(4+)
Safety and Handling

Safety and Handling

GHS H Statement
H314-H226-H336
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
May cause drowsiness or dizziness.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

SDS Documents