Learn More
Zirconium(IV) fluoride, low hafnium, 99%, Hf <0.1%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01154222
Description
Zirconium fluoride is used as a zirconium source in oxygen-sensitive applications such as metal production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zirconium(IV) fluoride | |
Low hafnium | |
White | |
100 g | |
MFCD00011307 | |
14,10175 | |
167.21 | |
Hf <0.1% | |
Moisture sensitive |
600°C (sublimation) | |
7783-64-4 | |
Powder | |
ZrF4 | |
UN3260 | |
Insoluble in water. | |
99% | |
1.59 | |
4.43 g/mL |
Chemical Identifiers
7783-64-4 | |
MFCD00011307 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 232-018-1
RTECSNumber : ZH7875000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only