Learn More
Zirconium(IV) fluoride, 98%, Thermo Scientific Chemicals
$168.14 - $816.52
Chemical Identifiers
CAS | 7783-64-4 |
---|---|
Molecular Formula | F4Zr |
MDL Number | MFCD00011307 |
Synonym | Zirconium tetrafluoride |
Description
Zirconium(IV) fluoride is used as a zirconium source in oxygen-sensitive applications such as metal production. Together with uranium salt, zirconium fluoride can be a component of fuel-coolant in molten salt reactors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-64-4 | |
F4Zr | |
MFCD00011307 | |
Zirconium tetrafluoride |
Specifications
600°C (sublimation) | |
White | |
50 g | |
ZrF4 | |
UN3260 | |
Zirconium tetrafluoride | |
167.21 | |
1.59 | |
4.43 g/mL |
7783-64-4 | |
Powder | |
F4Zr | |
MFCD00011307 | |
14,10175 | |
Slightly soluble in water; Soluble in hydrofluoric acid | |
98% | |
Moisture sensitive | |
Zirconium (IV) fluoride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 232-018-1
RTECSNumber : ZH7875000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only