missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium(IV) ethoxide, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 335450050

 View more versions of this product

Catalog No. AC335450050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zirconium(IV) ethoxide
18267-08-8
White
Authentic
Glass bottle
Zr(OC2H5)4
5 g
VCYRFVPNTBNCLW-UHFFFAOYSA-N
ethanolate;zirconium(4+)
122130910
99%
99%
200°C
235°C (7.0 Mbar)
98.5% min
C2H5OZr
MFCD00015654,MFCD00015654
zirconium iv ethoxide
[Zr+4].CC[O-]
136.28
271.46
Crystalline Powder and/or Chunks

Chemical Identifiers

18267-08-8
136.28
VCYRFVPNTBNCLW-UHFFFAOYSA-N
122130910
[Zr+4].CC[O-]
C2H5OZr
MFCD00015654,MFCD00015654
zirconium iv ethoxide
ethanolate;zirconium(4+)
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Call a POISON CENTER or doctor/physician

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only