Learn More
Zirconium(IV) ethoxide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 335450250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zirconium(IV) ethoxide | |
18267-08-8 | |
White | |
Authentic | |
Glass bottle | |
Zr(OC2H5)4 | |
25 g | |
VCYRFVPNTBNCLW-UHFFFAOYSA-N | |
ethanolate;zirconium(4+) | |
122130910 | |
99% |
99% | |
200°C | |
235°C (7.0 Mbar) | |
98.5% min | |
C2H5OZr | |
MFCD00015654,MFCD00015654 | |
zirconium iv ethoxide | |
[Zr+4].CC[O-] | |
136.28 | |
271.46 | |
Crystalline Powder and/or Chunks |
Chemical Identifiers
18267-08-8 | |
136.28 | |
VCYRFVPNTBNCLW-UHFFFAOYSA-N | |
122130910 | |
[Zr+4].CC[O-] |
C2H5OZr | |
MFCD00015654,MFCD00015654 | |
zirconium iv ethoxide | |
ethanolate;zirconium(4+) |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable solid.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Call a POISON CENTER or doctor/physician
GHS Signal Word: Danger
RUO – Research Use Only