Learn More
Zirconium(IV) chloride, Reactor Grade, 99.5+% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01210409
Description
Zirconium tetrachloride is commonly utilized as a Lewis acid catalyst in many organic synthesis reactions. It is an important source for organometallic complexes of zirconium. It is practiced in many areas like nuclear industry, metal industry, photography, glass, and ceramic industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zirconium(IV) chloride | |
10026-11-6 | |
10 g | |
ZrCl4 | |
Zirconium tetrachloride | |
233.03 | |
Moisture sensitive |
331° (sublimation) | |
Powder | |
>99.5% (metals basis) | |
MFCD00011306 | |
Soluble in alcohol,ether,hydrochloric acid. | |
Reactor | |
2.803 |
Chemical Identifiers
10026-11-6 | |
ZrCl4 | |
MFCD00011306 | |
Zirconium tetrachloride |
Safety and Handling
P234-P264b-P270-P280-P284-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P331-P363-P390-P501c
H290-H302-H314
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 233-058-2
RTECSNumber : ZH7175000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only