missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium(IV) Bromide, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 8613918

 View more versions of this product

Catalog No. AA8613918



Specifications

Specifications

Zirconium(IV) Bromide
99%
MFCD00049637
LSWWNKUULMMMIL-UHFFFAOYSA-J
tetrabromozirconium
83727
Moisture sensitive
13777-25-8
Br4Zr
zirconium tetrabromide, zrbr4, zirconium iv bromide, zirconium bromide zrbr4 , t-4, zirconium bromide zrbr4, zirconium bromide zrbr4 , t-4, zirconium bromide zrbr4 8ci 9ci, zirconium iv bromide, anhydrous trace metals basis
Br[Zr](Br)(Br)Br
410.84
410.86
50g

Chemical Identifiers

13777-25-8
410.84
LSWWNKUULMMMIL-UHFFFAOYSA-J
83727
Br[Zr](Br)(Br)Br
Br4Zr
MFCD00049637
zirconium tetrabromide, zrbr4, zirconium iv bromide, zirconium bromide zrbr4 , t-4, zirconium bromide zrbr4, zirconium bromide zrbr4 , t-4, zirconium bromide zrbr4 8ci 9ci, zirconium iv bromide, anhydrous trace metals basis
tetrabromozirconium
Safety and Handling

Safety and Handling

When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

Causes burns.

DOTInformation : DOT Class: 8

EINECSNumber : 237-417-4

SDS Documents