Learn More
Zirconium(IV) 2-ethylhexanoate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03917422
Description
Zirconium(IV) 2-ethylhexanoate is utilized as a metal organic precursor of zirconium for solution deposition and chemical vapor deposition of thin films containing zirconium especially zirconium oxide thin films. It is also used as yttria stabilized zirconia films for solid oxide fuel cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zirconium(IV) 2-ethylhexanoate | |
Amber | |
C32H60O8Zr | |
100 g | |
zirconium tetra 2-ethylhexanoate, zirconium 2-ethylhexanoate, 2-ethylhexanoate; zirconium 4+, zirconium iv 2-ethylhexanoate, acmc-1cfv3, zirconium octoate, 2-ethylhexanoate; zirconium +4 cation, zirconium iv 2-ethylhexanoate 25g, tetrakis 2-ethylhexanoic acid zirconium iv salt, zirconium 4+ tetrakis 2-ethylhexanoate | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zr+4] | |
664.048 | |
664.04 | |
Viscous Liquid |
2233-42-3 | |
>93°C (199°F) | |
MFCD00072684 | |
Moisture sensitive | |
OFYFURKXMHQOGG-UHFFFAOYSA-J | |
2-ethylhexanoate;zirconium(4+) | |
6451377 | |
97% |
Chemical Identifiers
2233-42-3 | |
664.048 | |
OFYFURKXMHQOGG-UHFFFAOYSA-J | |
6451377 | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Zr+4] |
C32H60O8Zr | |
MFCD00072684 | |
zirconium tetra 2-ethylhexanoate, zirconium 2-ethylhexanoate, 2-ethylhexanoate; zirconium 4+, zirconium iv 2-ethylhexanoate, acmc-1cfv3, zirconium octoate, 2-ethylhexanoate; zirconium +4 cation, zirconium iv 2-ethylhexanoate 25g, tetrakis 2-ethylhexanoic acid zirconium iv salt, zirconium 4+ tetrakis 2-ethylhexanoate | |
2-ethylhexanoate;zirconium(4+) |
Safety and Handling
GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
P201-P202-P210-P281-P308+P313-P370+P378q-P501c
H227-H361
EINECSNumber : 218-776-6
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only