missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium(IV) 2,4-pentanedionate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01406518

 View more versions of this product

Catalog No. AA1406518



Description

Description

Zirconium(IV) 2,4-pentanedionate is used as a cross-linking agent for oxygen-containing polymers. It is used as a catalyst in organic synthesis for the nitration of o-nitrotoluene using a dinitrogen pentoxide to get 2,4-dinitrotoluenes and 2,6-dinitrotoluenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zirconium(IV) 2,4-pentanedionate
191°C to 195°C
Weak
MFCD00000036
4-hydroxypent-3-en-2-one; zirconium
DFPGFWYWSAYLCW-UHFFFAOYSA-N
4-hydroxypent-3-en-2-one;zirconium
50912253
Crystals
17501-44-9
White
C20H28O8Zr
50 g
Soluble in water.
[Zr+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
487.66
487.66

Chemical Identifiers

17501-44-9
487.66
DFPGFWYWSAYLCW-UHFFFAOYSA-N
50912253
[Zr+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
C20H28O8Zr
MFCD00000036
4-hydroxypent-3-en-2-one; zirconium
4-hydroxypent-3-en-2-one;zirconium
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 241-510-5

RTECSNumber : ZH9280000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only