Learn More
Zirconium(IV) 2,4-pentanedionate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01406530
Description
Zirconium(IV) 2,4-pentanedionate is used as a cross-linking agent for oxygen-containing polymers. It is used as a catalyst in organic synthesis for the nitration of o-nitrotoluene using a dinitrogen pentoxide to get 2,4-dinitrotoluenes and 2,6-dinitrotoluenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zirconium(IV) 2,4-pentanedionate | |
191°C to 195°C | |
Weak | |
MFCD00000036 | |
4-hydroxypent-3-en-2-one; zirconium | |
DFPGFWYWSAYLCW-UHFFFAOYSA-N | |
4-hydroxypent-3-en-2-one;zirconium | |
50912253 | |
Crystals |
17501-44-9 | |
White | |
C20H28O8Zr | |
250 g | |
Soluble in water. | |
[Zr+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
487.66 | |
487.66 |
Chemical Identifiers
17501-44-9 | |
487.66 | |
DFPGFWYWSAYLCW-UHFFFAOYSA-N | |
50912253 | |
[Zr+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
C20H28O8Zr | |
MFCD00000036 | |
4-hydroxypent-3-en-2-one; zirconium | |
4-hydroxypent-3-en-2-one;zirconium |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 241-510-5
RTECSNumber : ZH9280000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only