Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium(IV) 1,1,1-trifluoro-2,4-pentanedionate, 98+%

$45.72 - $155.47

Chemical Identifiers

Chemical Name or Material Zirconium(IV) 1,1,1-trifluoro-2,4-pentanedionate
CAS 17499-68-2
Formula Weight 703.46
MDL Number MFCD00015317
Synonym Zirconium(IV) trifluoroacetylacetonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1927603

  SDS
alfa aesar™
1927603
1g Each for $45.72

AA1927606

  SDS
alfa aesar™
1927606
5g Each for $155.47

AA1927614

  SDS
alfa aesar™
1927614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 703.46
Chemical Name or Material Zirconium(IV) 1,1,1-trifluoro-2,4-pentanedionate
Physical Form Crystalline
Assay ≥98%
MDL Number MFCD00015317
Quantity 1g
CAS 17499-68-2
Boiling Point 235°C (dec.)
Synonym Zirconium(IV) trifluoroacetylacetonate
Solubility Information Soluble in toluene; Slightly soluble in THF and methylene chloride; Insoluble in water