missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium(IV) 1,1,1-trifluoro-2,4-pentanedionate, 98+%

Manufacturer:  Alfa Aesar™ 1927603

 View more versions of this product

Catalog No. AA1927603



Specifications

Specifications

Zirconium(IV) 1,1,1-trifluoro-2,4-pentanedionate
1g
C20H16F12O8Zr
zirconium iv tetrakis 1,1,1-trifluoro-4-oxo-2-pentene-2-olate
WYYHZWGGPPBCMA-YSGGVRKFSA-J
(Z)-1,1,1-trifluoro-4-oxopent-2-en-2-olate;(E)-1,1,1-trifluoro-4-oxopent-2-en-2-olate;zirconium(4+)
101644371
Crystalline
235°C (decomposition)
17499-68-2
MFCD00015317
Soluble in toluene; Slightly soluble in THF and methylene chloride; Insoluble in water
CC(=O)C=C(C(F)(F)F)[O-].CC(=O)C=C(C(F)(F)F)[O-].CC(=O)C=C(C(F)(F)F)[O-].CC(=O)C=C(C(F)(F)F)[O-].[Zr+4]
703.545
703.46
≥98%

Chemical Identifiers

17499-68-2
703.545
WYYHZWGGPPBCMA-YSGGVRKFSA-J
101644371
CC(=O)C=C(C(F)(F)F)[O-].CC(=O)C=C(C(F)(F)F)[O-].CC(=O)C=C(C(F)(F)F)[O-].CC(=O)C=C(C(F)(F)F)[O-].[Zr+4]
C20H16F12O8Zr
MFCD00015317
zirconium iv tetrakis 1,1,1-trifluoro-4-oxo-2-pentene-2-olate
(Z)-1,1,1-trifluoro-4-oxopent-2-en-2-olate;(E)-1,1,1-trifluoro-4-oxopent-2-en-2-olate;zirconium(4+)
SDS Documents