missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium hydride, 99%

Manufacturer:  Alfa Aesar™ 4720330

 View more versions of this product

Catalog No. AA4720330



Specifications

Specifications

Zirconium hydride
MFCD00148878
14,10176
99%
Odorless
250g
7704-99-6
UN1437
Powder
−325 Mesh
Moisture Sensitive

Chemical Identifiers

7704-99-6
MFCD00148878
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packaging Group: II; Proper Shipping Name: ZIRCONIUM HYDRIDE

TSCA : TSCA

SDS Documents