missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium foil, 1.0mm (0.039 in.) thick, annealed, 99.2% (metals basis excluding Hf), Hf 4.5% max

Manufacturer:  Alfa Aesar™ 10590FI

 View more versions of this product

Catalog No. AA10590FI



Specifications

Specifications

Zirconium foil
50 x 50mm
7440-67-7
Zr
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
[Zr]
Odorless
23995
Foil
≈16.3g/50x50mm
1.0mm (0.039 in.) thick, Annealed, Temper: soft
(metals basis excluding Hf), Hf 4.5% max.
MFCD00011303
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zirconium
91.224
CHEBI:33342
99.2%

Chemical Identifiers

7440-67-7
91.224
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
zirconium
Zr
MFCD00011303
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
CHEBI:33342
[Zr]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents