missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zirconium foil, 1.0mm (0.039in) thick, annealed, 99.2% (metals basis excluding Hf), Hf 4.5% max, Thermo Scientific Chemicals
$100.66 - $719.32
Chemical Identifiers
CAS | 7440-67-7 |
---|---|
Molecular Formula | Zr |
Molecular Weight (g/mol) | 91.22 |
MDL Number | MFCD00011303 MFCD00148878 |
InChI Key | QCWXUUIWCKQGHC-UHFFFAOYSA-N |
Synonym | zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil |
PubChem CID | 23995 |
ChEBI | CHEBI:33342 |
IUPAC Name | zirconium |
SMILES | [Zr] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA10590FI
|
Thermo Scientific Chemicals
010590FI |
50 x 50 mm |
Each for $100.66
|
|
|||||
AA10590GH
|
Thermo Scientific Chemicals
010590GH |
100 x 100 mm |
Each for $221.58
|
|
|||||
AA10590HP
|
Thermo Scientific Chemicals
010590HP |
100 x 500 mm |
Each for $719.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-67-7 | |
91.22 | |
QCWXUUIWCKQGHC-UHFFFAOYSA-N | |
23995 | |
zirconium |
Zr | |
MFCD00011303 MFCD00148878 | |
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil | |
CHEBI:33342 | |
[Zr] |
Specifications
7440-67-7 | |
MFCD00011303 MFCD00148878 | |
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil | |
[Zr] | |
91.22 | |
CHEBI:33342 | |
Odorless | |
Foil | |
(metals basis excluding Hf), Hf 4.5% max. |
Zr | |
50 x 50 mm | |
QCWXUUIWCKQGHC-UHFFFAOYSA-N | |
zirconium | |
23995 | |
99.2% | |
≈16.3g/50x50mm | |
Zirconium foil, 1.0mm (0.039 in.) thick, Annealed, Temper: soft |
Safety and Handling
EINECSNumber : 231-176-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only