missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium Foil, 0.25mm (0.01 in.) thick, annealed, 99.8% (Metals basis excluding Hf), Alfa Aesar™

Form: Approx.20.6g/100 x 125mm, Temper: soft

Manufacturer:  Alfa Aesar 10591GR

 View more versions of this product

Catalog No. AA10591GR



Specifications

Specifications

Zirconium
Annealed
Gray to silver foil
99.8% (Metals basis excluding Hf)
MFCD00011303
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zirconium
91.224
CHEBI:33342
Gray to Silver
125 x 500mm
7440-67-7
6.506g/cm3
Zr
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
[Zr]
Odorless
23995
Foil, 0.25mm (0.01 in.) thick

Chemical Identifiers

7440-67-7
91.224
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
zirconium
Zr
MFCD00011303
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
CHEBI:33342
[Zr]
Documents