missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium foil, 0.20mm (0.008 in.) thick, annealed, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 44752IB

 View more versions of this product

Catalog No. AA44752IB



Specifications

Specifications

Zirconium foil
125 x 2000mm
7440-67-7
Zr
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
Odorless
zirconium
23995
Foil
≈20.3g/100x125mm
0.20mm (0.008 in.) thick, Annealed, Temper: soft
(metals basis)
MFCD00011303
QCWXUUIWCKQGHC-UHFFFAOYSA-N
[Zr]
91.224
CHEBI:33342
99.8%

Chemical Identifiers

7440-67-7
91.224
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
zirconium
Zr
MFCD00011303
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
CHEBI:33342
[Zr]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents