missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium Foil, 0.127mm (0.005 in.) thick, annealed, 99.5% (Metals basis), Alfa Aesar™

Form: Approx.10.3g/100 x 125mm, Temper: soft

Manufacturer:  Alfa Aesar 10594GM

 View more versions of this product

Catalog No. AA10594GM



Specifications

Specifications

Zirconium
Annealed
Gray to silver foil
Zr
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
[Zr]
Odorless
23995
Foil, 0.127mm (0.005 in.) thick
100 x 125mm
7440-67-7
99.5% (Metals basis)
MFCD00011303
QCWXUUIWCKQGHC-UHFFFAOYSA-N
zirconium
91.224
CHEBI:33342
Gray to Silver

Chemical Identifiers

7440-67-7
91.224
QCWXUUIWCKQGHC-UHFFFAOYSA-N
23995
zirconium
Zr
MFCD00011303
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
CHEBI:33342
[Zr]
Documents