missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zirconium foil, 0.127mm (0.005in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Form: Approx.10.3g/100 x 125mm, Temper: soft
Supplier: Thermo Scientific Chemicals 010594GR
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gray to Silver | |
Annealed | |
Gray to silver foil | |
Zr | |
125 x 500 mm | |
QCWXUUIWCKQGHC-UHFFFAOYSA-N | |
zirconium | |
23995 | |
Odorless |
Zirconium | |
7440-67-7 | |
99.5% (Metals basis) | |
MFCD00011303 MFCD00148878 | |
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil | |
[Zr] | |
91.22 | |
CHEBI:33342 | |
Foil |
Chemical Identifiers
7440-67-7 | |
91.22 | |
QCWXUUIWCKQGHC-UHFFFAOYSA-N | |
23995 | |
zirconium |
Zr | |
MFCD00011303 MFCD00148878 | |
zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil | |
CHEBI:33342 | |
[Zr] |
Safety and Handling
EINECSNumber : 231-176-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only