Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium dinitrate oxide hydrate, Puratronic™, 99.994% (metals basis)

Chemical Identifiers

Chemical Name or Material Zirconium dinitrate oxide hydrate
CAS 14985-18-3
Purity Grade Puratronic™
Linear Formula ZrO(NO3)2·xH2O
Formula Weight 231.23 (Anhydrous)
MDL Number MFCD00149896
UN Number UN1477
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1297106

  SDS
alfa aesar™
1297106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1297114

  SDS
alfa aesar™
1297114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1297122

  SDS
alfa aesar™
1297122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 231.23 (Anhydrous)
MDL Number MFCD00149896
TSCA TSCA
Health Hazard 3 GHS P Statement
P221-P210-P220-P280-P305+P351+P338-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
Assay 99.994%
Purity Grade Puratronic™
Physical Form Powder
CAS 14985-18-3
Health Hazard 2 GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
Linear Formula ZrO(NO3)2·xH2O
Odor Odorless
Color White
Quantity 5g
Chemical Name or Material Zirconium dinitrate oxide hydrate
UN Number UN1477
Assay Percent Notes (metals basis)
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P280-P305+P351+P338-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.