missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zirconium Dichloride Oxide Hydrate, 99.9985% (Metal Basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1113530

 View more versions of this product

Catalog No. AA1113530



Specifications

Specifications

Zirconium Dichloride Oxide Hydrate
99.9985%, metal basis
MFCD00149897
VBTPZBJEHPOUOI-UHFFFAOYSA-N
oxozirconium;hydrate;dihydrochloride
16211499
Crystalline Aggregates
15461-27-5
Cl2H4O2Zr
cl2ozr.h2o
O.O=[Zr].Cl.Cl
198.154
178.13
250g

Chemical Identifiers

15461-27-5
198.154
VBTPZBJEHPOUOI-UHFFFAOYSA-N
16211499
O.O=[Zr].Cl.Cl
Cl2H4O2Zr
MFCD00149897
cl2ozr.h2o
oxozirconium;hydrate;dihydrochloride
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

Harmful if swallowed. Causes burns. Possible risk of irreversible effects

DOTInformation : DOT Class: 8

Documents