Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium carbide, 99.5% (metals basis excluding Hf), Hf< 200ppm

Chemical Identifiers

Chemical Name or Material Zirconium carbide
CAS 12070-14-3
Formula Weight 103.23
MDL Number MFCD00049640
UN Number UN3178
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3580818

  SDS
alfa aesar™
3580818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3580830

  SDS
alfa aesar™
3580830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 103.23
Chemical Name or Material Zirconium carbide
TSCA TSCA
Mesh Size −325 Mesh
Odor Odorless
Physical Form Powder
Assay 99.5%
Assay Percent Notes (metals basis excluding Hf), Hf< 200ppm
MDL Number MFCD00049640
CAS 12070-14-3
UN Number UN3178
Color Gray
Melting Point 3540°C
Quantity 50g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228
Flammable solid.
Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Density 6.73g/mL
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
RTECS Number ZH7155000
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

GHS H Statement
H228
Flammable solid.