missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Zirconium boride, 99.5% (metals basis excluding Hf)

Manufacturer:  Alfa Aesar™ 4703018

 View more versions of this product

Catalog No. AA4703018



Specifications

Specifications

Zirconium boride
99.5%
B2Zr
MFCD00064648
VWZIXVXBCBBRGP-UHFFFAOYSA-N
boron;zirconium
9812765
Powder
3000°C
Zirconium diboride
12045-64-6
(metals basis excluding Hf)
ZrB2
UN3178
[B].[B].[Zr]
112.844
112.84
15 Micron APS
50g

Chemical Identifiers

12045-64-6
112.844
VWZIXVXBCBBRGP-UHFFFAOYSA-N
boron;zirconium
B2Zr
MFCD00064648
9812765
[B].[B].[Zr]
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

RTECSNumber : ZH7150000

TSCA : TSCA

SDS Documents