Please login to your online account to display your discounted pricing

Zinterol hydrochloride, Tocris

Selective β2 agonist

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material Zinterol hydrochloride
Assay Percent Range >98%
Molecular Formula C19H26N2O4S
Formula Weight 414.95
Synonym N-[5-[2-[(1, 1-Dimethyl-2-phenylethyl)amino]-1-hydroxyethyl]-2-hydroxyphenyl]methanesulphonamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

105110

 
r & d systems
RDS105110
10mg Each for $214.12

105150

 
r & d systems
RDS105150
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 414.95
Quantity 10mg
Chemical Name or Material Zinterol hydrochloride
Assay Percent Range >98%
CAS 37000-20-7
Synonym N-[5-[2-[(1, 1-Dimethyl-2-phenylethyl)amino]-1-hydroxyethyl]-2-hydroxyphenyl]methanesulphonamide hydrochloride
Molecular Formula C19H26N2O4S
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C