Please login to your online account to display your discounted pricing

Zinterol hydrochloride, Tocris


Selective β2 agonist

Quantity 50mg

Manufacturer: r & d systems  RDS105150

 View more versions of this product

Catalog No. 105150

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Zinterol hydrochloride
CAS 37000-20-7
Recommended Storage Store at +4°C
Assay Percent Range >98%
Molecular Formula C19H26N2O4S
Formula Weight 414.95
Quantity 50mg
Synonym N-[5-[2-[(1, 1-Dimethyl-2-phenylethyl)amino]-1-hydroxyethyl]-2-hydroxyphenyl]methanesulphonamide hydrochloride

Specifications

Chemical Name or Material Zinterol hydrochloride
Assay Percent Range >98%
Molecular Formula C19H26N2O4S
Formula Weight 414.95
Synonym N-[5-[2-[(1, 1-Dimethyl-2-phenylethyl)amino]-1-hydroxyethyl]-2-hydroxyphenyl]methanesulphonamide hydrochloride
Accessories

Store at +4°C