Please login to your online account to display your discounted pricing

Zinc wire, 0.5mm (0.02in) dia, Puratronic™, 99.994% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Zinc wire
CAS 7440-66-6
Name Note 0.5mm (0.02 in.) dia, Puratronic™
MDL Number MFCD00011291
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA12054G5

  SDS
alfa aesar
12054G5
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5m
Chemical Name or Material Zinc wire
TSCA TSCA
Density ≈1.40g/m
Name Note 0.5mm (0.02 in.) dia, Puratronic™
Assay 99.994%
Assay Percent Notes (metals basis)
MDL Number MFCD00011291
CAS 7440-66-6
Odor Odorless