Please login to your online account to display your discounted pricing

Alfa Aesar™ Zinc tungsten oxide, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Zinc tungsten oxide
CAS 13597-56-3
Formula Weight 313.22
MDL Number MFCD00049633
Synonym Zinc tungstate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4023409

  SDS
alfa aesar™
4023409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4023418

  SDS
alfa aesar™
4023418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4023430

  SDS
alfa aesar™
4023430
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 313.22
Synonym Zinc tungstate
Health Hazard 1 Warning
Mesh Size −200 Mesh
TSCA TSCA
Health Hazard 3 GHS P Statement
P261-P304+P340-P312-P405-P403+P233-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99.9%
Physical Form Powder
Chemical Name or Material Zinc tungsten oxide
Assay Percent Notes (metals basis)
Health Hazard 2 GHS H Statement
H335
May cause respiratory irritation.
CAS 13597-56-3
Quantity 10g
MDL Number MFCD00049633
Safety and Handling

Warning

GHS P Statement
P261-P304+P340-P312-P405-P403+P233-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Store in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H335
May cause respiratory irritation.