missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Zinc tungsten oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04023418
Description
It is used as a pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Zinc tungsten oxide | |
Powder | |
(metals basis) | |
MFCD00049633 | |
Insoluble in water. | |
[O-][W](=O)(=O)[O-].[Zn+2] | |
313.216 | |
313.22 |
13597-56-3 | |
50 g | |
O4WZn | |
zinc tungsten oxide, zinc tungstate, zinkwolframat, wo4.zn, zinc 2+ tungstate, zinc, ion zn2+ tungstate | |
LJSXSLGFYGBEPA-UHFFFAOYSA-N | |
zinc;dioxido(dioxo)tungsten | |
22022519 | |
99.9% |
Chemical Identifiers
13597-56-3 | |
313.216 | |
LJSXSLGFYGBEPA-UHFFFAOYSA-N | |
22022519 | |
[O-][W](=O)(=O)[O-].[Zn+2] |
O4WZn | |
MFCD00049633 | |
zinc tungsten oxide, zinc tungstate, zinkwolframat, wo4.zn, zinc 2+ tungstate, zinc, ion zn2+ tungstate | |
zinc;dioxido(dioxo)tungsten |
Safety and Handling
GHS H Statement
H335
May cause respiratory irritation.
P261-P271-P304+P340-P312-P501c
H335
EINECSNumber : 237-053-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only