missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc tetrafluoroborate hydrate, Zn 18% min, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 08512430

 View more versions of this product

Catalog No. AA8512430



Description

Description

Zinc tetrafluoroborate hydrate is used as a precursor for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties. Further, it serves as a mild catalyst for epoxide ring opening with amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Zinc Tetrafluoroborate
Powder
B2F8Zn
zinc tetrafluoroborate hydrate, 2bf4.zn.h2o, zinc tetrafluoroborate-water 1/2/1, zinc 2+ hydrate ditetrafluoroborate, zinc 2+ ion hydrate ditetrafluoroborate, borate 1-,tetrafluoro-, zinc 2:1 , hexahydrate 9ci
BUMIVIYHDVNOQE-UHFFFAOYSA-N
zinc(2+) bis(tetrafluoroboranuide)
16212112
Hygroscopic
27860-83-9
250 g
MFCD00150371
Soluble in water and alcohol.
[Zn++].F[B-](F)(F)F.F[B-](F)(F)F
238.99
238.98 (Anhydrous)

Chemical Identifiers

27860-83-9
238.99
BUMIVIYHDVNOQE-UHFFFAOYSA-N
16212112
B2F8Zn
MFCD00150371
zinc tetrafluoroborate hydrate, 2bf4.zn.h2o, zinc tetrafluoroborate-water 1/2/1, zinc 2+ hydrate ditetrafluoroborate, zinc 2+ ion hydrate ditetrafluoroborate, borate 1-,tetrafluoro-, zinc 2:1 , hexahydrate 9ci
[Zn++].F[B-](F)(F)F.F[B-](F)(F)F
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : DOT Class: 8; Packaging Group: III

EINECSNumber : 237-534-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only