missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Zinc Tetrafluoroborate Hydrate, Zn <18%, Alfa Aesar™

Manufacturer:  Alfa Aesar 8512418

 View more versions of this product

Catalog No. AA8512418



Specifications

Specifications

Zinc Tetrafluoroborate
B2F8H2OZn
zinc tetrafluoroborate hydrate, 2bf4.zn.h2o, zinc tetrafluoroborate-water 1/2/1, zinc 2+ hydrate ditetrafluoroborate, zinc 2+ ion hydrate ditetrafluoroborate, borate 1-,tetrafluoro-, zinc 2:1 , hexahydrate 9ci
[B-](F)(F)(F)F.[B-](F)(F)(F)F.O.[Zn+2]
257.002
238.98 (anhydrous)
Hydrate
27860-83-9
MFCD00150371
WKVXCUNBQMAPGE-UHFFFAOYSA-N
zinc;ditetrafluoroborate;hydrate
16212112
Powder
50g

Chemical Identifiers

27860-83-9
257.002
WKVXCUNBQMAPGE-UHFFFAOYSA-N
16212112
[B-](F)(F)(F)F.[B-](F)(F)(F)F.O.[Zn+2]
B2F8H2OZn
MFCD00150371
zinc tetrafluoroborate hydrate, 2bf4.zn.h2o, zinc tetrafluoroborate-water 1/2/1, zinc 2+ hydrate ditetrafluoroborate, zinc 2+ ion hydrate ditetrafluoroborate, borate 1-,tetrafluoro-, zinc 2:1 , hexahydrate 9ci
zinc;ditetrafluoroborate;hydrate
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

Causes burns.

DOTInformation : DOT Class: 8

EINECSNumber : 237-534-0

SDS Documents